1-tert-Butylguanidine hydrochloride

Lieferant: Apollo Scientific

OR300901-1G
APOSOR300901-1GEA 0 EUR
APOSOR300901-1G
1-tert-Butylguanidine hydrochloride
1-tert-Butylguanidine hydrochloride
Formel: C₅H₁₃N₃·ClH
Molecular Weight: 151,638 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28122847
CAS-Nummer: 26536-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR