Pyrrolidine-1-carboxamidine hydrochloride

Lieferant: Apollo Scientific

OR300903-1G
APOSOR300903-1GEA 0 EUR
APOSOR300903-1G
Pyrrolidine-1-carboxamidine hydrochloride
Pyrrolidine-1-carboxamidine hydrochloride
Formel: C₅H₁₁N₃·ClH
Molecular Weight: 149,622 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25963774
CAS-Nummer: 49755-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR