(2,4-Dioxo-thiazolidin-5-yl)-acetic acid

Lieferant: Apollo Scientific

Synonyme: 2-(2,4-Dioxothiazolidin-5-yl)acetic acid

OR300906-1G OR300906-250MG OR300906-500MG
APOSOR300906-1GEA 221 EUR
APOSOR300906-1G APOSOR300906-250MG APOSOR300906-500MG
(2,4-Dioxo-thiazolidin-5-yl)-acetic acid
(2,4-Dioxothiazolidin-5-yl)acetic acid
Formel: C₅H₅NO₄S
Molecular Weight: 175.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01165868
CAS-Nummer: 875-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR