2-(Benzylamino)-2-phenylacetonitrile

Lieferant: Apollo Scientific

OR300908-1G OR300908-250MG OR300908-500MG OR300908-5G
APOSOR300908-1GEA 197 EUR
APOSOR300908-1G APOSOR300908-250MG APOSOR300908-500MG APOSOR300908-5G
2-(Benzylamino)-2-phenylacetonitrile
2-(Benzylamino)-2-phenylacetonitrile
Formel: C₁₅H₁₄N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00631398
CAS-Nummer: 32153-18-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR