6-Methoxy-4-hydroxyquinoline

Lieferant: Apollo Scientific

Synonyme: 6-Methoxyquinolin-4-ol

OR301003-5G
APOSOR301003-5GEA 82.8 EUR
APOSOR301003-5G
6-Methoxy-4-hydroxyquinoline
6-Methoxy-4-hydroxyquinoline
Formel: C₁₀H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00014662
CAS-Nummer: 23432-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR