2-Chloro-5-methoxybenzonitrile
Lieferant: Apollo Scientific
OR301005-1G
APOSOR301005-1GEA
21.9
EUR
APOSOR301005-1G
2-Chloro-5-methoxybenzonitrile
2-Chloro-5-methoxybenzonitrile
Formel:
C₈H₆ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD08061919 CAS-Nummer: 127667-00-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...