2-Bromimidazo[1,2-a]pyridin

Lieferant: Apollo Scientific
Achtung

OR301012-1G
APOSOR301012-1GEA 84.1 EUR
APOSOR301012-1G
2-Bromimidazo[1,2-a]pyridin
2-Bromimidazo[1,2-a]pyridin
Formel: C₇H₅BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09834778
CAS-Nummer: 112581-95-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR