5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5,6,7,8-Tetrahydro-1,2,4-triazolo[4,3-a]pyrazine Hydrochloride
OR301013-1G
APOSOR301013-1GEA
85.7
EUR
APOSOR301013-1G
5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride
5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride
Formel:
C₅H₉ClN₄ Molecular Weight: 160,61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08448159 CAS-Nummer: 837430-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...