3-Brom-o-kresol

Lieferant: Apollo Scientific
Achtung

OR301014-5G OR301014-25G
APOSOR301014-5GEA 72.5 EUR
APOSOR301014-5G APOSOR301014-25G
3-Brom-o-kresol
3-Brom-o-kresol
Formel: C₇H₇BrO
Lagertemperatur: Raumtemperatur
MDL: MFCD11100990
CAS-Nummer: 7766-23-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR