3-Amino-6,6-dimethyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole, N5-BOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 3-amino-6,6-dimethylpyrrolo[3,4-c]pyrazole-5(1H,4H,6H)-carboxylate, tert-Butyl 3-amino-4,6-dihydro-6,6-dimethylpyrrolo[3,4-c]pyrazole-5(1H)-carboxylate, 3-Amino-5-(tert-butoxycarbonyl)-6,6-dimethyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole

OR301022-1G
APOSOR301022-1GEA 119 EUR
APOSOR301022-1G
3-Amino-6,6-dimethyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole, N5-BOC protected
3-Amino-6,6-dimethyl-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole, N5-BOC protected
Formel: C₁₂H₂₀N₄O₂
Molecular Weight: 252,313 g/mol
Schmelzpunkt: 100…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08275051
CAS-Nummer: 398491-61-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR