1H-Pyrrolo[2,3-b]pyridin-5-ol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Hydroxy-7-azaindole

OR301023-25G
APOSOR301023-25GEA 168 EUR
APOSOR301023-25G
1H-Pyrrolo[2,3-b]pyridin-5-ol 97%
1H-Pyrrolo[2,3-b]pyridin-5-ol
Formel: C₇H₆N₂O
Molecular Weight: 134.14 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08272242
CAS-Nummer: 98549-88-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR