5-Methoxy-1,3-dihydro-2H-indol-2-on

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Methoxy-2-indolinone, 5-Methoxyindolin-2-one

OR301024-1G
APOSOR301024-1GEA 30.6 EUR
APOSOR301024-1G
5-Methoxy-1,3-dihydro-2H-indol-2-on
5-Methoxyoxindole
Formel: C9H9NO2
Molecular Weight: 163,1733 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01860251
CAS-Nummer: 7699-18-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR