4-Hydroxy-1-methyl-1H-pyrazole 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methyl-1H-pyrazol-4-ol
OR301025-250MG
APOSOR301025-250MGEA
43.5
EUR
APOSOR301025-250MG
4-Hydroxy-1-methyl-1H-pyrazole 98%
4-Hydroxy-1-methyl-1H-pyrazole
Formel:
C₄H₆N₂O Molecular Weight: 98,103 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22414500 CAS-Nummer: 78242-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...