4-Amino-5-chloropyrimidine

Lieferant: Apollo Scientific

OR301026-1G
APOSOR301026-1GEA 132 EUR
APOSOR301026-1G
4-Amino-5-chloropyrimidine
4-Amino-5-chloropyrimidine
Formel: C₄H₄ClN₃
Molecular Weight: 129,5500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00233964
CAS-Nummer: 101257-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR