3-(Aminomethyl)benzonitril 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Cyanobenzylamine
OR301027-5G
APOSOR301027-5GEA
144
EUR
APOSOR301027-5G
3-(Aminomethyl)benzonitril 98%
3-(Aminomethyl)benzonitril
Formel:
C₈H₈N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797832 CAS-Nummer: 10406-24-3 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...