Methyl 2-methyl-6-nitrobenzoate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 6-nitro-o-toluate, 2-(Methoxycarbonyl)-3-nitrotoluene, 2-(Methoxycarbonyl)-3-methylnitrobenzene

OR301028-5G OR301028-1G
APOSOR301028-5GEA 63.8 EUR
APOSOR301028-5G APOSOR301028-1G
Methyl 2-methyl-6-nitrobenzoate
Methyl 2-methyl-6-nitrobenzoate
Formel: C₉H₉NO₄
Molecular Weight: 195.172 g/mol
Schmelzpunkt: 50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11042453
CAS-Nummer: 61940-22-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR