4-Cbz-aminopiperidine

Lieferant: Apollo Scientific

Synonyme: 4-Benzyloxycarbonylaminopiperidine, 4-Cbz-Aminopiperidine

OR301035-5G
APOSOR301035-5GEA 55.1 EUR
APOSOR301035-5G
4-Cbz-aminopiperidine
4-Cbz-aminopiperidine
Formel: C₁₃H₁₈N₂O₂
Molecular Weight: 234.2942 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038571
CAS-Nummer: 182223-54-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR