4-Aminocyclopentene hydrochloride

Lieferant: Apollo Scientific
Achtung

OR301049-1G
APOSOR301049-1GEA 0 EUR
APOSOR301049-1G
4-Aminocyclopentene hydrochloride
4-Aminocyclopentene hydrochloride
Formel: C₅H₁₀ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD01630740
CAS-Nummer: 91469-55-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR