Methyl-3-(brommethyl)benzoat
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Methyl 3-(bromomethyl)benzoate
OR30105-5G
APOSOR30105-5GEA
21.9
EUR
APOSOR30105-5G
Methyl-3-(brommethyl)benzoat
Methyl-3-(brommethyl)benzoat
Formel:
C₉H₉BrO₂ Molecular Weight: 229,07 g/mol Schmelzpunkt: 43…46 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051437 CAS-Nummer: 1129-28-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...