4,5-Dihydro-1H-benzo[b]azepin-2(3H)-one

Lieferant: Apollo Scientific

Synonyme: 1,3,4,5-Tetrahydrobenzo[b]azepin-2-one

OR301054-5G
APOSOR301054-5GEA 94.4 EUR
APOSOR301054-5G
4,5-Dihydro-1H-benzo[b]azepin-2(3H)-one
4,5-Dihydro-1H-benzo[b]azepin-2(3H)-one
Formel: C₁₀H₁₁NO
Molecular Weight: 161,2004 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00024149
CAS-Nummer: 4424-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR