2-Methyl-5-iodopyridine

Lieferant: Apollo Scientific

Synonyme: 5-Iodo-2-methylpyridine

OR301057-5G
APOSOR301057-5GEA 484 EUR
APOSOR301057-5G
2-Methyl-5-iodopyridine
2-Methyl-5-iodopyridine
Formel: C₆H₆IN
Molecular Weight: 219.023 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07437990
CAS-Nummer: 695-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR