Pyrrolo[2,1-f][1,2,4]triazin-4(3H)-one
Lieferant: Apollo Scientific
OR301058-1G
APOSOR301058-1GEA
88.6
EUR
APOSOR301058-1G
Pyrrolo[2,1-f][1,2,4]triazin-4(3H)-one
Pyrrolo[2,1-f][1,2,4]triazin-4(3H)-one
Formel:
C₆H₅N₃O Molecular Weight: 135,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12923122 CAS-Nummer: 159326-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...