N-Benzyl-1,3-propandiamin

Lieferant: Apollo Scientific
Gefahr

OR301059-1G
APOSOR301059-1GEA 0 EUR
APOSOR301059-1G
N-Benzyl-1,3-propandiamin
N1-Benzylpropane-1,3-diamine
Formel: C₁₀H₁₆N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00274361
CAS-Nummer: 13910-48-0
UN: 2735
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR