2,4-Dichloro-5H-pyrrolo[3,2-d]pyrimidine 98%

Lieferant: Apollo Scientific
Achtung

OR301065-25G
APOSOR301065-25GEA 150 EUR
APOSOR301065-25G
2,4-Dichloro-5H-pyrrolo[3,2-d]pyrimidine 98%
2,4-Dichloro-5H-pyrrolo[3,2-d]pyrimidine
Formel: C₆H₃Cl₂N₃
Molecular Weight: 188.0141 g/mol
Schmelzpunkt: 229…231 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08059281
CAS-Nummer: 63200-54-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR