2-Brom-p-anisaldehyd 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-4-methoxybenzaldehyde

OR301068-25G OR301068-100G OR301068-5G
APOSOR301068-25GEA 84.1 EUR
APOSOR301068-25G APOSOR301068-100G APOSOR301068-5G
2-Brom-p-anisaldehyd 98%
2-Brom-p-anisaldehyd
Formel: C₈H₇BrO₂
Molecular Weight: 215.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09788636
CAS-Nummer: 43192-31-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR