2-Brom-5-chlorthiazol

Lieferant: Apollo Scientific
Achtung

OR301069-1G
APOSOR301069-1GEA 89.9 EUR
APOSOR301069-1G
2-Brom-5-chlorthiazol
2-Bromo-5-chlorothiazole
Formel: C₃HBrClNS
Molecular Weight: 198,4687 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09909626
CAS-Nummer: 16629-15-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR