2-Bromo-6-nitropyridine

Lieferant: Apollo Scientific

OR301070-1G
APOSOR301070-1GEA 111 EUR
APOSOR301070-1G
2-Bromo-6-nitropyridine
2-Bromo-6-nitropyridine
Formel: C₅H₃BrN₂O₂
Molecular Weight: 202,9935 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234012
CAS-Nummer: 21203-78-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR