3-Formyl-2-methoxypyridin

Lieferant: Apollo Scientific

Synonyme: 2-Methoxynicotinaldehyde, 2-Methoxy-3-pyridinecarboxaldehyde, 2-Methoxy-pyridine-3-carbaldehyde, 2-Methoxypyridine-3-carboxaldehyde

OR301072-5G
APOSOR301072-5GEA 74.1 EUR
APOSOR301072-5G
3-Formyl-2-methoxypyridin
3-Formyl-2-methoxypyridin
Formel: C₇H₇NO₂
Molecular Weight: 137,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115112
CAS-Nummer: 71255-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR