6-Bromo-2-hydroxyquinoxaline

Lieferant: Apollo Scientific

Synonyme: 6-Bromoquinoxalin-2(1H)-one

OR301106-1G
APOSOR301106-1GEA 104 EUR
APOSOR301106-1G
6-Bromo-2-hydroxyquinoxaline
6-Bromo-2-hydroxyquinoxaline
Formel: C₈H₅BrN₂O
Molecular Weight: 225.0421 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09834132
CAS-Nummer: 55687-34-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR