7-Chlorimidazo[4,5-b]pyridin
Lieferant: Apollo Scientific
Synonyme:
7-Chloro-1H-imidazo[4,5-b]pyridine
OR301113-1G
APOSOR301113-1GEA
104
EUR
APOSOR301113-1G
7-Chlorimidazo[4,5-b]pyridin
7-Chlorimidazo[4,5-b]pyridin
Formel:
C₆H₄ClN₃ Molecular Weight: 153,5691 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15143399 CAS-Nummer: 6980-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...