2-Methyl-2-propanyl-3-cyan-4-oxo-1-pyrrolidincarboxylat 97%

Lieferant: Apollo Scientific

Synonyme: 3-Cyano-4-oxopyrrolidine-1-carboxylic acid tert-Butyl ester, tert-Butyl 3-cyano-4-oxopyrrolidine-1-carboxylate, 1-Boc-3-cyano-4-pyrrolidone, tert-Butyl 3-Cyano-4-oxopyrrolidine-1-carboxylate, 1-tert-Butoxycarbonyl-3-cyano-4-pyrrolidone, 1-Boc-3-cyano-4-pyrrolidinone, 1-tert-Butoxycarbonyl-3-cyano-4-pyrrolidinone

OR301132-100G
APOSOR301132-100GEA 134 EUR
APOSOR301132-100G
2-Methyl-2-propanyl-3-cyan-4-oxo-1-pyrrolidincarboxylat 97%
tert-Butyl-3-cyano-4-oxopyrrolidine-1-carboxylate
Formel: C₁₀H₁₄N₂O₃
Molecular Weight: 210,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01318456
CAS-Nummer: 175463-32-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR