7-Bromo-1H-pyrrolo[3,2-c]pyridine

Lieferant: Apollo Scientific

OR301138-1G
APOSOR301138-1GEA 382 EUR
APOSOR301138-1G
7-Bromo-1H-pyrrolo[3,2-c]pyridine
7-Bromo-1H-pyrrolo[3,2-c]pyridine
Formel: C₇H₅BrN₂
Molecular Weight: 197,0320 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781153
CAS-Nummer: 902837-42-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR