3,4-Dichlor-N-methylanilin

Lieferant: Apollo Scientific

Synonyme: N1-Methyl-3,4-dichloroaniline

OR30114-5G
APOSOR30114-5GEA 0 EUR
APOSOR30114-5G
3,4-Dichlor-N-methylanilin
3,4-Dichlor-N-methylanilin
Formel: C₇H₇Cl₂N
Molecular Weight: 176.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052850
CAS-Nummer: 40750-59-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR