(1H-Indol-2-yl)methanamine
Lieferant: Apollo Scientific
OR301148-1G
APOSOR301148-1GEA
116
EUR
APOSOR301148-1G
(1H-Indol-2-yl)methanamine
(1H-Indol-2-yl)methanamine
Formel:
C₉H₁₀N₂ Molecular Weight: 146,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03422512 CAS-Nummer: 21109-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...