1-(4-(Bromomethyl)phenyl)ethanone
Lieferant: Apollo Scientific
OR301151-25G
APOSOR301151-25GEA
0
EUR
APOSOR301151-25G
1-(4-(Bromomethyl)phenyl)ethanone
1-(4-(Bromomethyl)phenyl)ethanone
Formel:
C₉H₉BrO Lagertemperatur: Raumtemperatur |
MDL:
MFCD10001463 CAS-Nummer: 51229-51-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...