2,4-Dichloro-5,7-dihydrofuro[3,4-d]pyrimidine

Lieferant: Apollo Scientific

OR301154-1G
APOSOR301154-1GEA 75.4 EUR
APOSOR301154-1G
2,4-Dichloro-5,7-dihydrofuro[3,4-d]pyrimidine
2,4-Dichloro-5,7-dihydrofuro[3,4-d]pyrimidine
Formel: C₆H₄Cl₂N₂O
Molecular Weight: 191,0200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13250060
CAS-Nummer: 848398-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR