2,4-Dichloro-6-methoxypyrimidine

Lieferant: Apollo Scientific
Achtung

OR301155-250MG
APOSOR301155-250MGEA 24.8 EUR
APOSOR301155-250MG
2,4-Dichloro-6-methoxypyrimidine
2,4-Dichloro-6-methoxypyrimidine
Formel: C₅H₄Cl₂N₂O
Molecular Weight: 179,0000 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00955635
CAS-Nummer: 43212-41-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR