2H-Thiopyran-3,5(4H,6H)-dione

Lieferant: Apollo Scientific

OR301158-1G
APOSOR301158-1GEA 632 EUR
APOSOR301158-1G
2H-Thiopyran-3,5(4H,6H)-dione
2H-Thiopyran-3,5(4H,6H)-dione
Formel: C₅H₆O₂S
Molecular Weight: 130,165 g/mol
Schmelzpunkt: 80…81 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06651188
CAS-Nummer: 6881-49-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR