3-(Benzyloxy)cyclobutanon 97%

Lieferant: Apollo Scientific
Achtung

OR301159-5G
APOSOR301159-5GEA 139 EUR
APOSOR301159-5G
3-(Benzyloxy)cyclobutanon 97%
3-(Benzyloxy)cyclobutanone
Formel: C11H12O2
Molecular Weight: 176.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09755991
CAS-Nummer: 30830-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR