3-Ethyl-2-iodtoluol
Lieferant: Apollo Scientific
|
Achtung
|
OR30116-10G
OR30116-25G
OR30116-1G
OR30116-5G
OR30116-50G
APOSOR30116-10GEA
111
EUR
APOSOR30116-10G
APOSOR30116-25G
APOSOR30116-1G
APOSOR30116-5G
APOSOR30116-50G
3-Ethyl-2-iodtoluol
3-Ethyl-2-iodtoluol
Formel:
C₉H₁₁I Siedepunkt: 118…120 °C/15mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052855 CAS-Nummer: 175277-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...