4-Amino-5-methylpyrimidine

Lieferant: Apollo Scientific

OR301165-1G
APOSOR301165-1GEA 65.4 EUR
APOSOR301165-1G
4-Amino-5-methylpyrimidine
4-Amino-5-methylpyrimidine
Formel: C₅H₇N₃
Molecular Weight: 109,1300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09909807
CAS-Nummer: 22433-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR