4-Bromo-2-methyl-1H-pyrrolo[2,3-b]pyridine 95%

Lieferant: Apollo Scientific
Achtung

OR301166-250MG
APOSOR301166-250MGEA 66.7 EUR
APOSOR301166-250MG
4-Bromo-2-methyl-1H-pyrrolo[2,3-b]pyridine 95%
4-Bromo-2-methyl-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₇BrN₂
Molecular Weight: 211,0586 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15529172
CAS-Nummer: 1014613-64-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR