4-Chlorofuro[3,2-d]pyrimidine

Lieferant: Apollo Scientific
Achtung

OR301170-250MG
APOSOR301170-250MGEA 258 EUR
APOSOR301170-250MG
4-Chlorofuro[3,2-d]pyrimidine
4-Chlorofuro[3,2-d]pyrimidine
Formel: C₆H₃ClN₂O
Molecular Weight: 154,5538 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09834944
CAS-Nummer: 655255-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR