tert-Butyl-3-oxoazepane-1-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Azepan-3-one, N-BOC protected, N-Boc-3-azacycloheptan-1-one

OR301179-1G
APOSOR301179-1GEA 91.5 EUR
APOSOR301179-1G
tert-Butyl-3-oxoazepane-1-carboxylate
tert-Butyl-3-oxoazepane-1-carboxylate
Formel: C₁₁H₁₉NO₃
Molecular Weight: 213,2735 g/mol
Siedepunkt: 90 °C/0,15mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD08443803
CAS-Nummer: 870842-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR