tert-Butyl-6-bromo-4-oxospiro[chroman-2,4'-piperidine]-1'-carboxylate
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 6-bromo-4-oxospiro[chroman-2,4'-piperidine]-1'-carboxylate
OR301180-1G
APOSOR301180-1GEA
203
EUR
APOSOR301180-1G
tert-Butyl-6-bromo-4-oxospiro[chroman-2,4'-piperidine]-1'-carboxylate
tert-Butyl-6-bromo-4-oxospiro[chroman-2,4'-piperidine]-1'-carboxylate
Formel:
C₁₈H₂₂BrNO₄ Molecular Weight: 396,2800 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05865132 CAS-Nummer: 690632-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...