2-(4-Iodophenyl)ethanol

Lieferant: Apollo Scientific

OR301185-5G
APOSOR301185-5GEA 104 EUR
APOSOR301185-5G
2-(4-Iodophenyl)ethanol
2-(4-Iodophenyl)ethanol
Formel: C₈H₉IO
Molecular Weight: 248,0600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173705
CAS-Nummer: 52914-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR