Ethyl-2-methylquinoline-3-carboxylate

Lieferant: Apollo Scientific
Gefahr

Synonyme: Ethyl 2-methylquinoline-3-carboxylate, 2-Methyl-quinoline-3-carboxylic acid ethyl ester

OR301187-1G OR301187-250MG
APOSOR301187-1GEA 81.2 EUR
APOSOR301187-1G APOSOR301187-250MG
Ethyl-2-methylquinoline-3-carboxylate
Ethyl-2-methylquinoline-3-carboxylate
Formel: C₁₃H₁₃NO₂
Molecular Weight: 215.25 g/mol
Schmelzpunkt: 70…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00195614
CAS-Nummer: 15785-08-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR