2-Methyl-5-pyrimidinboronsäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Methylpyrimidin-5-yl)boronic acid, 2-Methylpyrimidine-5-boronic acid

OR301199-5G
APOSOR301199-5GEA 178 EUR
APOSOR301199-5G
2-Methyl-5-pyrimidinboronsäure 96%
2-Methyl-5-pyrimidineboronic acid
Formel: C₅H₇BN₂O₂
Molecular Weight: 137,9323 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07375144
CAS-Nummer: 1034924-06-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR