4-Chloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine 97%

Lieferant: Apollo Scientific

OR301210-250MG
APOSOR301210-250MGEA 143 EUR
APOSOR301210-250MG
4-Chloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine 97%
4-Chloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine
Formel: C₇H₆ClN₃
Molecular Weight: 167,6000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09033903
CAS-Nummer: 35808-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR