(2-Methoxy-3-pyridinyl)methanol 97%
Lieferant: Apollo Scientific
Synonyme:
(2-Methoxypyridin-3-yl)methanol
OR301212-25G
APOSOR301212-25GEA
165
EUR
APOSOR301212-25G
(2-Methoxy-3-pyridinyl)methanol 97%
(2-Methoxy-3-pyridinyl)methanol
Formel:
C₇H₉NO₂ Molecular Weight: 139.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09880283 CAS-Nummer: 112197-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...